Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAAACAACAAAGGCGTGGGGGT[C/T]CCTGAGAAGGGTGGGAGGCACGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100740 MIM: 614469 MIM: 611481 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACHE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACHE - acetylcholinesterase (Cartwright blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000665.4 | 2121 | Intron | NP_000656.1 | |||
NM_001282449.1 | 2121 | Intron | NP_001269378.1 | |||
NM_001302621.1 | 2121 | UTR 3 | NP_001289550.1 | |||
NM_001302622.1 | 2121 | Intron | NP_001289551.1 | |||
NM_015831.2 | 2121 | UTR 3 | NP_056646.1 | |||
XM_006715995.2 | 2121 | UTR 3 | XP_006716058.1 | |||
XM_011516225.1 | 2121 | UTR 3 | XP_011514527.1 | |||
XM_011516226.1 | 2121 | UTR 3 | XP_011514528.1 | |||
XM_011516228.2 | 2121 | UTR 3 | XP_011514530.1 | |||
XM_011516229.2 | 2121 | UTR 3 | XP_011514531.1 | |||
XM_017012219.1 | 2121 | Intron | XP_016867708.1 | |||
XM_017012220.1 | 2121 | Intron | XP_016867709.1 |
SRRT - serrate, RNA effector molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFSP1 - UFM1 specific peptidase 1 (inactive) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |