Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCACGGACTTTTTTGAGTTGCC[A/G]GTAAGCAGGGTGTGGGTGCGTTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616861 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC12A9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC12A9 - solute carrier family 12 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267812.1 | Intron | NP_001254741.1 | ||||
NM_001267814.1 | Intron | NP_001254743.1 | ||||
NM_020246.3 | Intron | NP_064631.2 | ||||
XM_005250502.2 | Intron | XP_005250559.1 | ||||
XM_005250504.4 | Intron | XP_005250561.1 | ||||
XM_006716054.2 | Intron | XP_006716117.1 | ||||
XM_006716055.2 | Intron | XP_006716118.1 | ||||
XM_011516414.1 | Intron | XP_011514716.1 |
Set Membership: |
HapMap |