Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTTTCTTCACTGATTCCAGATT[A/C]ATATACTTTTCCACTGTTTTTCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614133 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEPACAM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HEPACAM2 - HEPACAM family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039372.2 | 1531 | UTR 3 | NP_001034461.1 | |||
NM_001288804.1 | 1531 | UTR 3 | NP_001275733.1 | |||
NM_001288810.1 | 1531 | UTR 3 | NP_001275739.1 | |||
NM_198151.2 | 1531 | UTR 3 | NP_937794.1 | |||
XM_011516000.2 | 1531 | UTR 3 | XP_011514302.1 | |||
XM_011516001.1 | 1531 | UTR 3 | XP_011514303.1 |