Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCACAAAATTTACAATACACTTT[C/T]TACAATTTAAAGTAATTTTTCCAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600610 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABPB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABPB1 - GA binding protein transcription factor beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320910.1 | Intron | NP_001307839.1 | ||||
NM_001320915.1 | Intron | NP_001307844.1 | ||||
NM_002041.4 | Intron | NP_002032.2 | ||||
NM_005254.5 | Intron | NP_005245.2 | ||||
NM_016654.4 | Intron | NP_057738.1 | ||||
NM_016655.4 | Intron | NP_057739.1 | ||||
NM_181427.3 | Intron | NP_852092.1 | ||||
XM_005254274.3 | Intron | XP_005254331.1 | ||||
XM_011521426.2 | Intron | XP_011519728.1 | ||||
XM_017022053.1 | Intron | XP_016877542.1 | ||||
XM_017022054.1 | Intron | XP_016877543.1 |