Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACCAACCAAGATATCATTGTTT[C/T]GATGTTTAAAGGAGCAGAATATGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603158 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP50 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP50 - ubiquitin specific peptidase 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP8 - ubiquitin specific peptidase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128610.2 | Intron | NP_001122082.1 | ||||
NM_001283049.1 | Intron | NP_001269978.1 | ||||
NM_005154.4 | Intron | NP_005145.3 | ||||
XM_006720761.3 | Intron | XP_006720824.1 | ||||
XM_006720762.3 | Intron | XP_006720825.1 | ||||
XM_011522193.2 | Intron | XP_011520495.1 | ||||
XM_017022718.1 | Intron | XP_016878207.1 | ||||
XM_017022719.1 | Intron | XP_016878208.1 | ||||
XM_017022720.1 | Intron | XP_016878209.1 | ||||
XM_017022721.1 | Intron | XP_016878210.1 | ||||
XM_017022722.1 | Intron | XP_016878211.1 |
Set Membership: |
HapMap |