Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGTTTAAATAACAGTAATTAT[C/T]TATCTCTTTCGTAGATCACCACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606902 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYGO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PYGO1 - pygopus family PHD finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015617.2 | 3027 | UTR 3 | NP_056432.1 | |||
XM_005254300.4 | 3027 | UTR 3 | XP_005254357.1 | |||
XM_011521446.2 | 3027 | UTR 3 | XP_011519748.1 |