Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTAATAGTATGAACTGTAGTGTT[C/T]TGGTACTTGGTTCATACAATTACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300469 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAGEC3 - MAGE family member C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138702.1 | Intron | NP_619647.1 | ||||
NM_177456.2 | Intron | NP_803251.1 | ||||
XM_011531267.2 | Intron | XP_011529569.1 | ||||
XM_017029265.1 | Intron | XP_016884754.1 |
Set Membership: |
HapMap |