Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATGCAGCATGATAGTACTATG[A/G]CAGTATACCAAGATAATACATTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605332 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLHL1 - kelch like family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286725.1 | Intron | NP_001273654.1 | ||||
NM_020866.2 | Intron | NP_065917.1 | ||||
XM_017020678.1 | Intron | XP_016876167.1 | ||||
XM_017020679.1 | Intron | XP_016876168.1 |
Set Membership: |
HapMap |