Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCCAAACTCTGGTCACCTTCT[C/T]ACTAGTCCTCAGAACCCAGCATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605873 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNK10 - potassium two pore domain channel subfamily K member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021161.4 | 6394 | UTR 3 | NP_066984.1 | |||
NM_138317.2 | 6394 | UTR 3 | NP_612190.1 | |||
NM_138318.2 | 6394 | UTR 3 | NP_612191.1 | |||
XM_011536840.2 | 6394 | UTR 3 | XP_011535142.1 |
LOC105370612 - uncharacterized LOC105370612 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |