Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGACTACATATATTAAATTTGGA[A/C]ATTCAAAGCATTAATAGAACAGAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TC2N PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TC2N - tandem C2 domains, nuclear | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128595.2 | 3652 | UTR 3 | NP_001122067.1 | |||
NM_001128596.2 | 3652 | UTR 3 | NP_001122068.1 | |||
NM_001289134.1 | 3652 | UTR 3 | NP_001276063.1 | |||
NM_152332.5 | 3652 | UTR 3 | NP_689545.1 |