Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCATGGTTTTGAAACACATTC[C/T]CTAAAAACCCTTCGATCCTGCTGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604580 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBLN5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBLN5 - fibulin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006329.3 | 2424 | UTR 3 | NP_006320.2 | |||
XM_005267267.3 | 2424 | UTR 3 | XP_005267324.1 | |||
XM_011536356.1 | 2424 | UTR 3 | XP_011534658.1 | |||
XM_011536357.1 | 2424 | UTR 3 | XP_011534659.1 | |||
XM_011536358.1 | 2424 | UTR 3 | XP_011534660.1 | |||
XM_017020929.1 | 2424 | UTR 3 | XP_016876418.1 |
TC2N - tandem C2 domains, nuclear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |