Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAATCTCTAAGAAATCTTGATCA[C/T]GTTTCCTGAATAAATTATGTCTTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602634 MIM: 612534 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAJB9 - DnaJ heat shock protein family (Hsp40) member B9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THAP5 - THAP domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130475.2 | Intron | NP_001123947.1 | ||||
NM_001287598.1 | Intron | NP_001274527.1 | ||||
NM_001287599.1 | Intron | NP_001274528.1 | ||||
NM_001287601.1 | Intron | NP_001274530.1 | ||||
NM_182529.3 | Intron | NP_872335.2 |