Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTCTTGCACCAGGTCCCTGCT[A/G]TATTTAGTGCCTCCAAGAATGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602378 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNM2 - dynamin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005360.2 | Intron | NP_001005360.1 | ||||
NM_001005361.2 | Intron | NP_001005361.1 | ||||
NM_001005362.2 | Intron | NP_001005362.1 | ||||
NM_001190716.1 | Intron | NP_001177645.1 | ||||
NM_004945.3 | Intron | NP_004936.2 |
Set Membership: |
HapMap |