Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606945 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDLR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LDLR - low density lipoprotein receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000527.4 | Intron | NP_000518.1 | ||||
NM_001195798.1 | Intron | NP_001182727.1 | ||||
NM_001195799.1 | Intron | NP_001182728.1 | ||||
NM_001195800.1 | Intron | NP_001182729.1 | ||||
NM_001195803.1 | Intron | NP_001182732.1 | ||||
XM_011528010.1 | Intron | XP_011526312.1 |
Set Membership: |
HapMap |