Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACGTACCCTGCCTTCATTCAT[C/T]TGTCCAACGCAGACATGTTGATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611840 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABHD8 - abhydrolase domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDA1 - DET1 and DDB1 associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024050.5 | Intron | NP_076955.1 |
MRPL34 - mitochondrial ribosomal protein L34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |