Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTTTTAGAAATGAGCTCAGGAT[A/G]TTTATAGATAAAAGAGATTTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612969 MIM: 601473 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIGD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TIGD7 - tigger transposable element derived 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF75A - zinc finger protein 75a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302109.1 | Intron | NP_001289038.1 | ||||
NM_001324041.1 | Intron | NP_001310970.1 | ||||
NM_001324085.1 | Intron | NP_001311014.1 | ||||
NM_153028.3 | Intron | NP_694573.1 | ||||
XM_011522646.2 | Intron | XP_011520948.1 | ||||
XM_011522647.2 | Intron | XP_011520949.1 | ||||
XM_017023642.1 | Intron | XP_016879131.1 | ||||
XM_017023643.1 | Intron | XP_016879132.1 | ||||
XM_017023644.1 | Intron | XP_016879133.1 | ||||
XM_017023645.1 | Intron | XP_016879134.1 |
Set Membership: |
HapMap |