Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGGTCAGGTAGCTGTAGGTCTT[A/T]GAAATGGCATCAAAGGTGGCCTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603843 MIM: 603624 MIM: 605915 | ||||||||||||||||||||
Literature Links: |
NDUFB10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFB10 - NADH:ubiquinone oxidoreductase subunit B10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004548.2 | Intron | NP_004539.1 |
RNF151 - ring finger protein 151 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL3L - ribosomal protein L3 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS2 - ribosomal protein S2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002952.3 | Intron | NP_002943.2 |
SNHG9 - small nucleolar RNA host gene 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA10 - small nucleolar RNA, H/ACA box 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA64 - small nucleolar RNA, H/ACA box 64 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA78 - small nucleolar RNA, H/ACA box 78 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBL3 - transducin beta like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |