Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTCCACCACCACCAGGTGCCGC[G/A]GGCCCAGGGCCCGGAACAGCTTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602727 | ||||||||||||||||||||
Literature Links: |
CCDC154 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC154 - coiled-coil domain containing 154 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLCN7 - chloride voltage-gated channel 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114331.2 | 2335 | Missense Mutation | CCG,CTG | P,L 742 | NP_001107803.1 | |
NM_001287.5 | 2335 | Missense Mutation | CCG,CTG | P,L 766 | NP_001278.1 | |
XM_011522354.1 | 2335 | Missense Mutation | CCG,CTG | P,L 708 | XP_011520656.1 |