Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTGGAAACCCTCTTTTTCATAC[C/T]GTTGGGCACCCACATACTCGTGCGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602844 MIM: 603816 MIM: 608012 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGDIG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGDIG - Rho GDP dissociation inhibitor gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AXIN1 - axin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003502.3 | Intron | NP_003493.1 | ||||
NM_181050.2 | Intron | NP_851393.1 | ||||
XM_011522682.2 | Intron | XP_011520984.1 | ||||
XM_011522683.2 | Intron | XP_011520985.1 | ||||
XM_011522684.2 | Intron | XP_011520986.1 | ||||
XM_011522686.1 | Intron | XP_011520988.1 | ||||
XM_017023743.1 | Intron | XP_016879232.1 | ||||
XM_017023744.1 | Intron | XP_016879233.1 | ||||
XM_017023745.1 | Intron | XP_016879234.1 | ||||
XM_017023746.1 | Intron | XP_016879235.1 | ||||
XM_017023747.1 | Intron | XP_016879236.1 | ||||
XM_017023748.1 | Intron | XP_016879237.1 |
PDIA2 - protein disulfide isomerase family A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |