Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGGGACTTCTCGCCTGCTTCTC[A/G]AGACACACTTCAGAAAATTCACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613560 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C9orf50 - chromosome 9 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199350.3 | Intron | NP_955382.3 | ||||
XM_011518655.1 | Intron | XP_011516957.1 | ||||
XM_011518656.1 | Intron | XP_011516958.1 | ||||
XM_011518657.2 | Intron | XP_011516959.1 | ||||
XM_011518658.2 | Intron | XP_011516960.1 | ||||
XM_011518659.1 | Intron | XP_011516961.1 | ||||
XM_011518660.1 | Intron | XP_011516962.1 | ||||
XM_011518661.2 | Intron | XP_011516963.1 | ||||
XM_011518662.2 | Intron | XP_011516964.1 | ||||
XM_011518663.1 | Intron | XP_011516965.1 | ||||
XM_017014708.1 | Intron | XP_016870197.1 |
NTMT1 - N-terminal Xaa-Pro-Lys N-methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286796.1 | Intron | NP_001273725.1 | ||||
NM_001286797.1 | Intron | NP_001273726.1 | ||||
NM_001286798.1 | Intron | NP_001273727.1 | ||||
NM_001286799.1 | Intron | NP_001273728.1 | ||||
NM_001286800.1 | Intron | NP_001273729.1 | ||||
NM_001286801.1 | Intron | NP_001273730.1 | ||||
NM_001286802.1 | Intron | NP_001273731.1 | ||||
NM_001286803.1 | Intron | NP_001273732.1 | ||||
NM_014064.3 | Intron | NP_054783.2 | ||||
XM_005251939.3 | Intron | XP_005251996.1 | ||||
XM_017014642.1 | Intron | XP_016870131.1 |
Set Membership: |
HapMap |