Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGAATAGCTAACATTTATAGA[C/G]GAATTTATTTTGTTTTACAAAGCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615027 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM60A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM60A - family with sequence similarity 60 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135811.1 | Intron | NP_001129283.1 | ||||
NM_001135812.1 | Intron | NP_001129284.1 | ||||
NM_021238.2 | Intron | NP_067061.1 | ||||
XM_011520803.2 | Intron | XP_011519105.1 | ||||
XM_017019758.1 | Intron | XP_016875247.1 | ||||
XM_017019759.1 | Intron | XP_016875248.1 | ||||
XM_017019760.1 | Intron | XP_016875249.1 | ||||
XM_017019761.1 | Intron | XP_016875250.1 | ||||
XM_017019762.1 | Intron | XP_016875251.1 |