Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612505 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABCA8 - ATP binding cassette subfamily A member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288985.1 | Intron | NP_001275914.1 | ||||
NM_001288986.1 | Intron | NP_001275915.1 | ||||
NM_007168.3 | Intron | NP_009099.1 | ||||
XM_005256938.2 | Intron | XP_005256995.1 | ||||
XM_005256940.4 | Intron | XP_005256997.1 | ||||
XM_011524191.2 | Intron | XP_011522493.1 | ||||
XM_011524192.2 | Intron | XP_011522494.1 |
Set Membership: |
HapMap |