Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGAGAGGTTGTGTCCAGGTCTC[A/C]GTGGTTTTCTTGGACACAACTATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604057 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HS3ST3A1 - heparan sulfate-glucosamine 3-sulfotransferase 3A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006042.2 | 2863 | UTR 3 | NP_006033.1 | |||
XM_011524114.2 | 2863 | UTR 3 | XP_011522416.1 | |||
XM_017025480.1 | 2863 | Intron | XP_016880969.1 |
Set Membership: |
HapMap |