Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTGGGAAATTGAGAGCATAGAC[A/G]TGAAGGAAATCAACTGAAAAAAGGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604057 | |||||||||||||||||||||||
Literature Links: |
HS3ST3A1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HS3ST3A1 - heparan sulfate-glucosamine 3-sulfotransferase 3A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006042.2 | 3951 | UTR 3 | NP_006033.1 | |||
XM_011524114.2 | 3951 | UTR 3 | XP_011522416.1 | |||
XM_017025480.1 | 3951 | Intron | XP_016880969.1 |
Set Membership: |
HapMap |