Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTATTTATATGAAGAAACCTCTC[C/T]TAAGCCACACTCCAATCACCCCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604334 MIM: 616463 | ||||||||||||||||||||
Literature Links: |
LOC101928000 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101928000 - uncharacterized LOC101928000 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP6 - ubiquitin specific peptidase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304284.1 | Intron | NP_001291213.1 | ||||
NM_004505.3 | Intron | NP_004496.2 | ||||
XM_011524050.1 | Intron | XP_011522352.1 | ||||
XM_011524051.2 | Intron | XP_011522353.1 | ||||
XM_011524052.2 | Intron | XP_011522354.1 | ||||
XM_011524053.2 | Intron | XP_011522355.1 | ||||
XM_011524054.2 | Intron | XP_011522356.1 | ||||
XM_011524055.2 | Intron | XP_011522357.1 | ||||
XM_011524056.2 | Intron | XP_011522358.1 | ||||
XM_011524057.2 | Intron | XP_011522359.1 | ||||
XM_011524058.2 | Intron | XP_011522360.1 | ||||
XM_011524059.2 | Intron | XP_011522361.1 | ||||
XM_017025290.1 | Intron | XP_016880779.1 |
ZNF232 - zinc finger protein 232 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320952.1 | Intron | NP_001307881.1 | ||||
NM_001320953.1 | Intron | NP_001307882.1 | ||||
NM_001320954.1 | Intron | NP_001307883.1 | ||||
NM_001320955.1 | Intron | NP_001307884.1 | ||||
NM_014519.3 | Intron | NP_055334.2 | ||||
XM_011524006.2 | Intron | XP_011522308.1 | ||||
XM_017025021.1 | Intron | XP_016880510.1 | ||||
XM_017025022.1 | Intron | XP_016880511.1 | ||||
XM_017025023.1 | Intron | XP_016880512.1 |