Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCAGCTCTGGGTGACGAGCAC[C/G]AGCCAAAGAGGCAGGATCTGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602076 MIM: 607066 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRPV1 - transient receptor potential cation channel subfamily V member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018727.5 | Intron | NP_061197.4 | ||||
NM_080704.3 | Intron | NP_542435.2 | ||||
NM_080705.3 | Intron | NP_542436.2 | ||||
NM_080706.3 | Intron | NP_542437.2 |
TRPV3 - transient receptor potential cation channel subfamily V member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |