Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTAAAACAACATGAATGGTAAA[A/C]GCAATTTTTTGTAACATTAGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605205 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADCY10 - adenylate cyclase 10, soluble | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167749.2 | 6520 | UTR 3 | NP_001161221.1 | |||
NM_001297772.1 | 6520 | UTR 3 | NP_001284701.1 | |||
NM_018417.5 | 6520 | UTR 3 | NP_060887.2 | |||
XM_005245330.4 | 6520 | Intron | XP_005245387.1 | |||
XM_006711449.3 | 6520 | UTR 3 | XP_006711512.1 | |||
XM_011509760.2 | 6520 | UTR 3 | XP_011508062.1 | |||
XM_011509762.2 | 6520 | UTR 3 | XP_011508064.1 | |||
XM_011509763.2 | 6520 | UTR 3 | XP_011508065.1 | |||
XM_011509766.2 | 6520 | Intron | XP_011508068.1 | |||
XM_017001778.1 | 6520 | Intron | XP_016857267.1 |