Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAAAATTCAAAAAAATACAAA[A/G]AAGTACCTACATGGTTACCACTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606033 MIM: 603395 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR2K PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POLR2K - RNA polymerase II subunit K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPAG1 - sperm associated antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003114.4 | Intron | NP_003105.2 | ||||
NM_172218.2 | Intron | NP_757367.1 | ||||
XM_011517240.2 | Intron | XP_011515542.1 | ||||
XM_011517241.2 | Intron | XP_011515543.1 | ||||
XM_011517242.2 | Intron | XP_011515544.1 | ||||
XM_011517243.2 | Intron | XP_011515545.1 | ||||
XM_011517245.2 | Intron | XP_011515547.1 | ||||
XM_017013754.1 | Intron | XP_016869243.1 | ||||
XM_017013755.1 | Intron | XP_016869244.1 |