Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACCAGGACCACCAACAGAGCCA[C/G]GGTGCAAACAATGTCAGTTGAATAA
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 607187 | ||||||||||||||||||||||||||
Literature Links: |
ST3GAL1 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ST3GAL1 - ST3 beta-galactoside alpha-2,3-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003033.3 | 3796 | Intron | NP_003024.1 | |||
NM_173344.2 | 3796 | Intron | NP_775479.1 | |||
XM_005251025.4 | 3796 | UTR 3 | XP_005251082.1 | |||
XM_006716617.1 | 3796 | UTR 3 | XP_006716680.1 | |||
XM_011517225.1 | 3796 | UTR 3 | XP_011515527.1 | |||
XM_017013736.1 | 3796 | UTR 3 | XP_016869225.1 | |||
XM_017013737.1 | 3796 | UTR 3 | XP_016869226.1 |
Set Membership: |
HapMap |