Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGTATTTCGATTTTCCATCTCCC[C/T]AGGCACGGTTGTCTTGAGAGGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605215 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf71 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C7orf71 - chromosome 7 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145531.2 | Intron | NP_001139003.1 | ||||
NM_001302625.1 | Intron | NP_001289554.1 | ||||
XM_017012010.1 | Intron | XP_016867499.1 | ||||
XM_017012011.1 | Intron | XP_016867500.1 | ||||
XM_017012012.1 | Intron | XP_016867501.1 | ||||
XM_017012013.1 | Intron | XP_016867502.1 | ||||
XM_017012014.1 | Intron | XP_016867503.1 |
SKAP2 - src kinase associated phosphoprotein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |