Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGCTTCTAAGTCGGAAAAGGT[C/T]GTGGCTTTAGCAAATTCAGTTTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607897 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSI2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MSI2 - musashi RNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322250.1 | Intron | NP_001309179.1 | ||||
NM_001322251.1 | Intron | NP_001309180.1 | ||||
NM_138962.3 | Intron | NP_620412.1 | ||||
NM_170721.1 | Intron | NP_733839.1 | ||||
XM_005257014.3 | Intron | XP_005257071.1 | ||||
XM_005257015.3 | Intron | XP_005257072.1 | ||||
XM_011524286.1 | Intron | XP_011522588.1 | ||||
XM_011524287.2 | Intron | XP_011522589.1 | ||||
XM_017024147.1 | Intron | XP_016879636.1 | ||||
XM_017024148.1 | Intron | XP_016879637.1 | ||||
XM_017024149.1 | Intron | XP_016879638.1 |
Set Membership: |
HapMap |