Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCCCTGGAGCATCCCACCAGC[C/G]CGGTGTGTGTGGCTGGCGTGGAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607934 MIM: 606755 | ||||||||||||||||||||
Literature Links: |
PADI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PADI1 - peptidyl arginine deiminase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PADI3 - peptidyl arginine deiminase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016233.2 | 89 | Missense Mutation | CCG,GCG | P,A 17 | NP_057317.2 | |
XM_011541571.2 | 89 | Intron | XP_011539873.1 | |||
XM_011541572.2 | 89 | Missense Mutation | CCG,GCG | P,A 17 | XP_011539874.1 | |
XM_017001463.1 | 89 | Intron | XP_016856952.1 |