Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACAGAGCCTGTATGCAGAGTGG[C/T]GCGAGTGTCTGGCCTGAGGAAAACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604886 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EMC8 - ER membrane protein complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142288.1 | Intron | NP_001135760.1 | ||||
NM_006067.4 | Intron | NP_006058.1 | ||||
XM_011522813.2 | Intron | XP_011521115.1 | ||||
XM_017022867.1 | Intron | XP_016878356.1 |