Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTATGGGAATATAGTGGTTAAGA[A/T]AACAGCTTCTCCCTTCATGGAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613438 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCHO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCHO2 - FCH domain only 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146032.1 | Intron | NP_001139504.1 | ||||
NM_138782.2 | Intron | NP_620137.2 | ||||
XM_017009016.1 | Intron | XP_016864505.1 | ||||
XM_017009017.1 | Intron | XP_016864506.1 | ||||
XM_017009018.1 | Intron | XP_016864507.1 | ||||
XM_017009019.1 | Intron | XP_016864508.1 | ||||
XM_017009020.1 | Intron | XP_016864509.1 | ||||
XM_017009021.1 | Intron | XP_016864510.1 | ||||
XM_017009022.1 | Intron | XP_016864511.1 | ||||
XM_017009023.1 | Intron | XP_016864512.1 |
Set Membership: |
HapMap |