Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTGATTTATGGACTGGTGGTC[A/G]GTGCAATAAGGTGTATACCCACTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612087 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC2A - C-type lectin domain family 2 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130711.1 | Intron | NP_001124183.1 | ||||
NM_207375.2 | Intron | NP_997258.1 | ||||
XM_011520654.2 | Intron | XP_011518956.1 | ||||
XM_011520656.2 | Intron | XP_011518958.1 | ||||
XM_011520657.2 | Intron | XP_011518959.1 |
KLRF2 - killer cell lectin like receptor F2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190765.1 | Intron | NP_001177694.1 |