Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAAAATATCCAGTGGCTCTTTT[A/C]AAAAACTTACTCATTATGGGAGCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615077 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D30 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBC1D30 - TBC1 domain family member 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015279.1 | Intron | NP_056094.1 | ||||
XM_011538078.2 | Intron | XP_011536380.1 | ||||
XM_017019083.1 | Intron | XP_016874572.1 | ||||
XM_017019084.1 | Intron | XP_016874573.1 | ||||
XM_017019085.1 | Intron | XP_016874574.1 | ||||
XM_017019086.1 | Intron | XP_016874575.1 | ||||
XM_017019087.1 | Intron | XP_016874576.1 |
Set Membership: |
HapMap |