Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACATTATTAATCTCCTGTAATT[C/T]AGCTCTGTCTTGGAGAACTGGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609229 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUDT4 - nudix hydrolase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301022.1 | Intron | NP_001287951.1 | ||||
NM_001301023.1 | Intron | NP_001287952.1 | ||||
NM_001301024.1 | Intron | NP_001287953.1 | ||||
NM_019094.5 | Intron | NP_061967.3 | ||||
NM_199040.3 | Intron | NP_950241.1 | ||||
XM_011537794.2 | Intron | XP_011536096.1 | ||||
XM_011537795.2 | Intron | XP_011536097.1 | ||||
XM_011537796.2 | Intron | XP_011536098.1 |
Set Membership: |
HapMap |