Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGGTTATATGTGAGCATCTGTG[C/T]TGTCCACTGCAACTAGACATGGGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176914 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1CC PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PPP1CC - protein phosphatase 1 catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244974.1 | Intron | NP_001231903.1 | ||||
NM_002710.3 | Intron | NP_002701.1 | ||||
XM_011538504.2 | Intron | XP_011536806.1 | ||||
XM_011538505.2 | Intron | XP_011536807.1 |
Set Membership: |
HapMap |