Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAATTGCTAGCTGGGACTGGACT[A/G]TAGCAGGAAGGGTGATGCTGATAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603766 MIM: 191343 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTIF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTIF2 - mitochondrial translational initiation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005369.1 | Intron | NP_001005369.1 | ||||
NM_001321001.1 | Intron | NP_001307930.1 | ||||
NM_001321002.1 | Intron | NP_001307931.1 | ||||
NM_001321003.1 | Intron | NP_001307932.1 | ||||
NM_001321004.1 | Intron | NP_001307933.1 | ||||
NM_001321005.1 | Intron | NP_001307934.1 | ||||
NM_002453.2 | Intron | NP_002444.2 | ||||
XM_005264335.3 | Intron | XP_005264392.1 | ||||
XM_011532871.2 | Intron | XP_011531173.1 | ||||
XM_017004162.1 | Intron | XP_016859651.1 | ||||
XM_017004163.1 | Intron | XP_016859652.1 | ||||
XM_017004164.1 | Intron | XP_016859653.1 | ||||
XM_017004165.1 | Intron | XP_016859654.1 | ||||
XM_017004166.1 | Intron | XP_016859655.1 | ||||
XM_017004167.1 | Intron | XP_016859656.1 |
RPS27A - ribosomal protein S27a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |