Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGGCCAGTCCTCCAGGCCGTG[A/G]TCAGTTCCAAGTTACTGTCTTCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609715 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TREML2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TREML2 - triggering receptor expressed on myeloid cells like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024807.3 | 1882 | UTR 3 | NP_079083.2 | |||
XM_011514917.2 | 1882 | UTR 3 | XP_011513219.1 |