Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGTCTGAAGCTATGTCCTAGA[C/T]GGTCCACTGTGTCACATGGCAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610774 MIM: 606817 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNPY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNPY3 - canopy FGF signaling regulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTCRA - pre T-cell antigen receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243168.1 | Intron | NP_001230097.1 | ||||
NM_001243169.1 | Intron | NP_001230098.1 | ||||
NM_001243170.1 | Intron | NP_001230099.1 | ||||
NM_138296.2 | Intron | NP_612153.2 |