Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTATTGTTGCTCTTCTAGGCC[C/G]ACGTTACACCTCCTACCTCCTCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607489 MIM: 600589 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CUL9 - cullin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015089.3 | 262 | Intron | NP_055904.1 | |||
XM_011514422.1 | 262 | Intron | XP_011512724.1 | |||
XM_011514423.2 | 262 | Intron | XP_011512725.1 | |||
XM_011514424.1 | 262 | Intron | XP_011512726.1 | |||
XM_011514425.1 | 262 | Intron | XP_011512727.1 | |||
XM_011514426.1 | 262 | Intron | XP_011512728.1 | |||
XM_011514427.1 | 262 | Intron | XP_011512729.1 | |||
XM_011514428.1 | 262 | Intron | XP_011512730.1 | |||
XM_011514429.1 | 262 | Intron | XP_011512731.1 | |||
XM_011514430.1 | 262 | Intron | XP_011512732.1 | |||
XM_017010589.1 | 262 | UTR 5 | XP_016866078.1 | |||
XM_017010590.1 | 262 | Intron | XP_016866079.1 |
SRF - serum response factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |