Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAACAGACATTTTGCTGCTGCTA[A/C]AAAGTAAGTATTTTTTTTTAGTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611821 | ||||||||||||||||||||
Literature Links: |
MRPL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL1 - mitochondrial ribosomal protein L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020236.3 | 338 | Missense Mutation | AAA,ACA | K,T 47 | NP_064621.3 | |
XM_011532198.2 | 338 | Missense Mutation | AAA,ACA | K,T 47 | XP_011530500.1 | |
XM_011532199.1 | 338 | Missense Mutation | AAA,ACA | K,T 47 | XP_011530501.1 |