Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTAGTCTGTATCTTCTGCTCTG[G/T]ATCTGTGTCTGTGTGTTAAGGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611574 | ||||||||||||||||||||
Literature Links: |
SGMS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SGMS2 - sphingomyelin synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136257.1 | Intron | NP_001129729.1 | ||||
NM_001136258.1 | Intron | NP_001129730.1 | ||||
NM_152621.5 | Intron | NP_689834.1 | ||||
XM_011531698.2 | Intron | XP_011530000.1 | ||||
XM_011531699.2 | Intron | XP_011530001.1 | ||||
XM_011531700.2 | Intron | XP_011530002.1 | ||||
XM_011531701.2 | Intron | XP_011530003.1 | ||||
XM_011531702.2 | Intron | XP_011530004.1 | ||||
XM_017007839.1 | Intron | XP_016863328.1 | ||||
XM_017007840.1 | Intron | XP_016863329.1 | ||||
XM_017007841.1 | Intron | XP_016863330.1 |