Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTATTATAGGCATCAGCAACCT[A/G]TCAGAAGAGAAATAGTTTTCCTTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615406 MIM: 611514 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG12-AS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GNG12-AS1 - GNG12 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WLS - wntless Wnt ligand secretion mediator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002292.3 | Intron | NP_001002292.3 | ||||
NM_001193334.1 | Intron | NP_001180263.1 | ||||
NM_024911.6 | Intron | NP_079187.3 | ||||
XM_011542191.2 | Intron | XP_011540493.1 | ||||
XM_011542192.2 | Intron | XP_011540494.1 | ||||
XM_017002390.1 | Intron | XP_016857879.1 |