Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTAGTCTTTTCTTGATGTCAAC[A/C]AAGTCTTTAAACCCCACAGCACTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163905 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HMGB1 - high mobility group box 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313892.1 | 3868 | UTR 3 | NP_001300821.1 | |||
NM_001313893.1 | 3868 | UTR 3 | NP_001300822.1 | |||
NM_002128.5 | 3868 | UTR 3 | NP_002119.1 |