Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCTAAGTATTTCCTTCTCTGTG[A/C]ACATCACTTAGGAGCTACGTCAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614795 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGPAT4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AGPAT4 - 1-acylglycerol-3-phosphate O-acyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020133.2 | 5193 | UTR 3 | NP_064518.1 | |||
XM_005267053.3 | 5193 | UTR 3 | XP_005267110.1 | |||
XM_006715514.3 | 5193 | UTR 3 | XP_006715577.1 | |||
XM_017011059.1 | 5193 | UTR 3 | XP_016866548.1 |
Set Membership: |
HapMap |