Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTAACTGCATTCATTTTTCATT[C/T]CCATGGACTCTCATCTCATCTTTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614825 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECT2L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ECT2L - epithelial cell transforming 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REPS1 - RALBP1 associated Eps domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128617.2 | Intron | NP_001122089.1 | ||||
NM_001286611.1 | Intron | NP_001273540.1 | ||||
NM_001286612.1 | Intron | NP_001273541.1 | ||||
NM_031922.4 | Intron | NP_114128.3 | ||||
XM_005267177.3 | Intron | XP_005267234.1 | ||||
XM_005267178.4 | Intron | XP_005267235.1 | ||||
XM_005267179.3 | Intron | XP_005267236.1 | ||||
XM_011536202.2 | Intron | XP_011534504.1 | ||||
XM_017011387.1 | Intron | XP_016866876.1 | ||||
XM_017011388.1 | Intron | XP_016866877.1 | ||||
XM_017011389.1 | Intron | XP_016866878.1 |
Set Membership: |
HapMap |