Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGGAGGATGGCCAAGGTCCCG[A/G]AGCTGGAAGACACCTTCCTGCAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612659 | ||||||||||||||||||||
Literature Links: |
RFX6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RFX6 - regulatory factor X6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173560.3 | 79 | Missense Mutation | AAG,GAG | K,E 6 | NP_775831.2 | |
XM_011535589.1 | 79 | Missense Mutation | AAG,GAG | K,E 6 | XP_011533891.1 | |
XM_017010477.1 | 79 | Intron | XP_016865966.1 |