Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCAACAGATAAAGAGTTCTAT[C/T]TGTCTTCTACGCGGGAAAATCTATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603461 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDC16 - cell division cycle 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005266206.1 | 581 | Silent Mutation | ATC,ATT | I,I 132 | XP_005266263.1 | |
XM_006719992.1 | 581 | Silent Mutation | ATC,ATT | I,I 131 | XP_006720055.1 | |
XM_006719993.1 | 581 | Silent Mutation | ATC,ATT | I,I 38 | XP_006720056.1 | |
XM_011534851.1 | 581 | Silent Mutation | ATC,ATT | I,I 132 | XP_011533153.1 | |
XM_011534853.1 | 581 | Silent Mutation | ATC,ATT | I,I 132 | XP_011533155.1 | |
XM_017020830.1 | 581 | Silent Mutation | ATC,ATT | I,I 132 | XP_016876319.1 | |
XM_017020831.1 | 581 | Silent Mutation | ATC,ATT | I,I 132 | XP_016876320.1 | |
XM_017020832.1 | 581 | Silent Mutation | ATC,ATT | I,I 38 | XP_016876321.1 | |
XM_017020833.1 | 581 | Silent Mutation | ATC,ATT | I,I 38 | XP_016876322.1 |
MIR548AR - microRNA 548ar | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |